Laboratory reactor systems, LR-2.ST the High Performer
Supplier: IKA
20013405
IKAA20013405EA 29310 SGD
IKAA20013405
Laboratory reactor systems, LR-2.ST the High Performer
Laboratory Reactor Systems
Modular laboratory reactor for the optimisation and reproduction of various chemical reactions, mixing and homogenisation processes on a lab scale. The system is characterised by the agitator mounting, which allows a safe transfer of the higher motor torque.
  • Suitable for vacuum operation
  • Solvent- and temperature-resistant perfluoro elastomer (FFPM) seals come into contact with the sample
  • Infinitely variable speed
  • Torque trend display for measuring changes in viscosity
  • Microprocessor-controlled speed regulation, enables steady speed, also under load
More Product Information