Heating bath, HB digital
Supplier: IKA
20016335
IKAA20016335EA 4320 SGD
IKAA20016335
Heating bath, HB digital
Baths Water Baths, Unstirred
Compact and ergonomic heating safety bath for all tempering media.
  • Maximum media temperature: 180 °C
  • Ergonomic handles
  • Key button to lock heating bath temperature
  • Clear, black-and-white display
  • Safety operating modes A, B, C
  • Permanent set-point/ actual temperature display
  • USB interface
Permissible ambient temperature 5 to 40 °C with 80% permissible relative humidity and frequency, 50/60 Hz.
Certifications: Protection class according to DIN EN 60529: IP 21, class designation acc. DIN 12876: II