Thermoshaker with heating and cooling, Matrix Delta Plus
Supplier: IKA
20100517 20100529
IKAA20100517EA 7110 SGD
IKAA20100517 IKAA20100529
Thermoshaker with heating and cooling, Matrix Delta Plus
Shakers Shaking Incubators
The IKA Matrix Delta Plus has heating and cooling functionality and numerous user-friendly features.
  • Large, clear display with a convenient menu structure allows for intuitive operation
  • Programmable ramps allow for automatic execution of heating and heating tasks
  • Secure, quick lock for fast and easy change of inserts and attachments as well as automatic attachment detection
  • Aluminium die-cast housing ensures secure positioning, excellent stability and everyday robustness
  • RS232 and USB interfaces
Matrix Delta Plus units heat and cool to ‒30 °C belowMore Product Information