Laboratory software, Labworldsoft 6
Supplier: IKA
20017366 20019397 20101872
IKAA20017366EA 32510 SGD
IKAA20017366 IKAA20019397 IKAA20101872
Laboratory software, Labworldsoft 6
Software Laboratory Software
Labworldsoft 6 is a laboratory software to control lab devices, automate processes, and to document your measurements and results. The software provides access to a huge library of different laboratory devices.
  • Easy recording of various physical parameters
  • Function modules to create time triggered or event triggered configurations
  • Communication through RS232, USB, Bluetooth or network
  • Supports formats .csv, .xslx and ASCII
  • Compatibility with Windows 7, 8 and 10
Labworldsoft does not integrate only IKA laboratory devices, but supports several otherMore Product Information