Magnetic stirrer, Maxi MR 1 digital IKAMAG®
Supplier: IKA
2621802 2621803 25004061 25004062
IKAA2621802EA 0 SGD
IKAA2621802 IKAA2621803 IKAA25004061 IKAA25004062
Magnetic stirrer, Maxi MR 1 digital IKAMAG®
Stirrers Magnetic Stirrers
Powerful high capacity magnetic stirrer with digital LED display.
  • Infinitely variable speed
  • Non-locking motor
  • Continuous operation or timer mode from 0 to 56 min
IP Protection Class according to DIN EN 60529: IP 21.