Reciprocating shaker, HS 501 digital
Supplier: IKA
2527000 2527003 2527032 25004443
IKAA2527000EA 8790 SGD
IKAA2527000 IKAA2527003 IKAA2527032 IKAA25004443
Reciprocating shaker, HS 501 digital
Shakers Reciprocating Shakers
Low profile laboratory shaker with digital display and a large platform. Continuous operation is guaranteed even under extreme loads. Suitable for vessels which need to be laid flat, for example separating funnels. A variety of different vessels can be accommodated by choosing from the range of optional holding attachments.
  • Infinitely variable speed control
  • Continuous operation or timer operation from 0 to 56 min
  • Electronic speed control
  • Can be used at 5 to 50 °C (80% relative humidity), in an incubator or cold room
IP protection class according toMore Product Information