Orbital shaker, KS 501 digital
Supplier: IKA
2526400 2526403 2526432 25004451
IKAA2526400EA 8790 SGD
IKAA2526400 IKAA2526403 IKAA2526432 IKAA25004451
Orbital shaker, KS 501 digital
Shakers Orbital Shakers
Low profile laboratory shaker with digital display which has a large mounting surface. Continuous operation is guaranteed even under extreme loads. Ideal for larger vessels of 250 ml and over, e.g. round bottomed flasks, Erlenmeyer flasks, culture flasks and bottles. Variety of different vessels can be accommodated by choosing from the range of optional holding attachments (not included with shaker).
  • Infinitely variable speed control continuous operation or timer operation from 0 to 55 min
  • Can be used at 5 to 50 °C (80% relative humidity), in an incubator
More Product Information