Magnetic hotplate stirrer, IKA Plate RCT digital
Supplier: IKA
25005221
IKAA25005221EA 4260 SGD
IKAA25005221
Magnetic hotplate stirrer, IKA Plate RCT digital
Stirrers Magnetic Hotplate Stirrers
The IKA Plate (RCT digital) magnetic hotplate stirrer has an aluminium alloy plate, suitable for optimum heat transfer. It features a glass housing surface for optimum cleaning and chemical resistance. The unit features a strong motor for a high range of speeds, integrated timer and counter function supports the monitoring of sensitive chemical reactions.
  • SmartTemp® hot surface warning to protect against burns
  • Connector for electronic temperature control (via optional PT1000 probe)
  • Regulation of the plate can be optimised by regular firmware updates
More Product Information