Magnetic hotplate stirrer, RCT basic
Supplier: IKA
25006455
IKAA25006455EA 3650 SGD
IKAA25006455
Magnetic hotplate stirrer, RCT basic
Stirrers Magnetic Hotplate Stirrers
The RCT basic magnetic stirrer provides reliability, an exceptionally long product lifetime and the highest safety standards. RCT basic is suitable for stirring tasks up to 20 L (H₂O) and reaches temperatures up to 310 °C. With the connection option for an external temperature sensor (PT1000.60), the temperature can be measured and controlled directly in the reaction medium. Thanks to insulation of the aluminium heating plate, maintenance-free EC motor and electronic switching power supply, RCT basic features excellent energy efficiency as well as reduced self-heating of the heating plateMore Product Information