Torque-measuring stirrer, STARVISC 200-2.5 control
Supplier: IKA
25006518
IKAA25006518EA 20240 SGD
IKAA25006518
Torque-measuring stirrer, STARVISC 200-2.5 control
Stirrers Overhead Stirrers
The STARVISC 200-2.5 control stirrer measures viscosity and displays it. The result can be read in real-time on the display. It has a broad range of applications.
  • High-precision and trend torque measurement
  • A viscosity calculation can be carried out immediately via a user friendly menu
  • Removable control unit and TFT display
  • Stepless speed control
  • Even highly viscous substances can be intensively stirred using the powerful STARVISC stirrer
This is particularly helpful during product development, clearly indicates while running research programmes as toMore Product Information