Microplate shaker, MTS 2/4 digital
Supplier: IKA
3208002 3208003
IKAA3208002EA 4250 SGD
IKAA3208002 IKAA3208003
Microplate shaker, MTS 2/4 digital
Shakers Microplate Shakers
Microplate shaker with electronic control keeps the set speed constant via rated/actual speed comparison. Supplied with attachment but without microtiter plate.
  • Holds two or four microplates
  • Digital timer with alarm allows a process time up to 99 minutes or continuous mode
  • Electronic speed control
  • Ideal for use under controlled conditions, can be used at 5 to 40 °C, in an incubator or CO₂ incubator (up to 80% RH)
IP protection class according to DIN EN 60529: IP 21
Certifications: IP protection class according to DIN EN 60529: IP 21