Vortex mixer, VORTEX Genius 3
Supplier: IKA
3340002 3340003
IKAA3340002EA 1460 SGD
IKAA3340002 IKAA3340003
Vortex mixer, VORTEX Genius 3
Shakers Vortex Shakers
Vortex mixer with three interchangeable attachments and various inserts for different applications. Microcentrifuge tubes, microtitre plates, even round bottomed or 250 ml Erlenmeyer flasks can be attached by means of the specially designed strap. Attachments click securely onto the unit in any position. The unit is stable at high speeds and sturdy with coated, zinc die-casting.
  • Compact design
  • Wide speed range, infinitely adjustable
  • Touch or continuous mode
  • Can be used at 5 to 40 °C (80% relative humidity) in an incubator or cold room
IP protectionMore Product Information