Electronic contact thermometers, ETS-D5/ ETS-D6
Supplier: IKA
3378000
IKAA3378000EA 1360 SGD
IKAA3378000
Electronic contact thermometers, ETS-D5/ ETS-D6
Thermometers Digital Thermometers
Ensure perfect temperature control without overshooting the set temperature, even in the case of rapid heating. With optimised PID control and RESET function. Suitable for use with all magnetic stirrers with contact thermometer bushing according to DIN 12878, class 2 (e.g. IKA®, Heidolph and Corning with adapter AD-C1).
  • Three modes of operation guarantee optimum adjustment to the working method
  • Operating mode A: Suitable for work with varying parameters (from −50 to +450 °C). Adjustable safety temperature
  • Operating mode B: Suitable for series operation
More Product Information