Magnetic hotplate stirrer, RET basic IKAMAG® safety control
Supplier: IKA
3622003 3622001 3622002
IKAA3622003EA 0 SGD
IKAA3622003 IKAA3622001 IKAA3622002
Magnetic hotplate stirrer, RET basic IKAMAG® safety control
Stirrers Magnetic Hotplate Stirrers
Magnetic hotplate stirrer with extremely fast heating times and integrated temperature control, suitable for unsupervised operation. Units have bushing according to DIN 12878 for connecting a contact thermometer, e.g. ETS-D5, enabling precise temperature control. Features include high magnetic adhesion, electronic speed control and digital error code display.
  • Exact temperature and speed setting via digital display, even when switched off
  • Safety temperature limit that has been set is displayed digitally
  • Increased safety due to improved heat control
More Product Information