Homogeniser, T 25 digital ULTRA-TURRAX®
Supplier: IKA
3725003 3725002 3725001
IKAA3725003EA 0 SGD
IKAA3725003 IKAA3725002 IKAA3725001
Homogeniser, T 25 digital ULTRA-TURRAX®
Homogenisers Homogenisers Bench
Stand model, volume range 1 to 2000 ml. The T 25 homogeniser is ideal for applications such as homogenising wastewater samples, for the use in laboratory reactors, dispersion tasks under vacuum/pressure and sample preparation in medical diagnostics.
  • Digital display for precise monitoring of set and actual speeds
  • Wide selection of dispersing tools to suit your application
  • Established rotor/stator configurations give consistent results
  • Motor protection against overload
IP protection class according to DIN EN 60529: IP 20.
Certifications: IPMore Product Information