Homogeniser, T 10 basic ULTRA-TURRAX®
Supplier: IKA
3737002 3737003
IKAA3737002EA 5630 SGD
IKAA3737002 IKAA3737003
Homogeniser, T 10 basic ULTRA-TURRAX®
Homogenisers Homogenisers Handheld
Handheld or stand model, volume range 0,5 to 100 ml. A wide speed range allows you to work at high circumferential speeds even with small rotor diameters.
  • Quick-release coupling makes changing the dispersing elements easy
  • Excellent speed stability with various media
  • Disposable plastic dispersing elements are available in two sizes
  • These are ideal for PCR analysis
IP protection class according to DIN EN 60529: IP 30
Certifications: IP protection class according to DIN EN 60529: IP 30
Delivery information: The homogeniser is supplied with an emptyMore Product Information