Electronic overhead stirrers, EUROSTAR 200 digital, EUROSTAR 200 control, EUROSTAR 200 P4 control
Supplier: IKA
3992003 3992002 3990003 4090003 4090002 3990002
IKAA3992003EA 0 SGD
IKAA3992003 IKAA3992002 IKAA3990003 IKAA4090003 IKAA4090002 IKAA3990002
Electronic overhead stirrers, EUROSTAR 200 digital, EUROSTAR 200 control, EUROSTAR 200 P4 control
Stirrers Overhead Stirrers
Extremely powerful laboratory stirrers for highly viscous applications and intensive mixing of quantities up to 100 L (H₂O). Units automatically adjust the speed through microprocessor controlled technology within the speed range of speed range of 6 to 2000 min⁻¹ or 4 to 530 min⁻¹ (P4 models), all models have two speed ranges. Safety circuits ensure automatic cut off in anti-stall or overload conditions. The actual shaft speed and preset speed are constantly monitored and variations are adjusted automatically. This guarantees a constant speed even with changes in sample viscosities.