Digital dry block heaters
Supplier: IKA
4025102 4025302 4025402
IKAA4025102EA 3470 SGD
IKAA4025102 IKAA4025302 IKAA4025402
Digital dry block heaters
Block Heaters Digital Block Heaters
Digital block heaters which hold from one up to four aluminum blocks, provide precise temperature control in small vessels. The PT1000 temperature sensor allows temperature control directly in the sample vessel. It is used with PCR tubes, PCR strips, Greiner tubes, microplates and cuvettes. The block heaters are ideal for melting and boiling point determination, enzyme reactions, incubation and activation of cultures, immunoassays, DNA denaturation, culture media tests, coagulation tests, blood/urea nitrogen determinations and in situ hybridisations.
  • Stepless adjustable temperature
More Product Information