Vortex mixer, VORTEX 1
Supplier: IKA
4047700
IKAA4047700EA 643 SGD
IKAA4047700
Vortex mixer, VORTEX 1
Shakers Vortex Shakers
Test tube mixer with touch function and infinitely adjustable speed from 1000 to 2800 min⁻¹. Ideal for mixing small samples. Upper casing and test tube holder is made from inert plastic, the bottom of the unit is coated, zinc die-casting.
  • Small, compact and reliable
  • Designed for small containers up to 30 mm Ø, such as test tubes and centrifuge tubes
  • Excellent mixing action
IP protection class according to DIN EN 60529: IP 40
Delivery information: Supplied with 12 V power pack.