Mechanical overhead stirrer, RW 47 digital
Supplier: IKA
4050000
IKAA4050000EA 17970 SGD
IKAA4050000
Mechanical overhead stirrer, RW 47 digital
Stirrers Overhead Stirrers
Extremely powerful, mechanically controlled laboratory stirrer designed for highly viscous applications for quantities up to 200 L. Unit has a robust, ergonomic design with constant power drive, suitable for intensive mixing for use in laboratories and pilot plants. The stirrer is equipped with protection to avoid overheating of the motor by means of self-locking temperature limiter. Additionally, to secure bowls, an optional stirring shaft protector and clamping fixture are available.
  • Digital speed display
  • Infinitely adjustable speed in two speed ranges
More Product Information
Order Now
SPECIFICATIONS
Model RW 47 digital
Max. stirring capacity H₂O 200 L
Environmentally Preferable
Max. viscosity 100000 mPas
Speed range I 57 - 275 min⁻¹
Speed range II 275 - 1300 min⁻¹
Max. torque 3000 Ncm
Chuck range 3 - 16 mm
Power - motor input/output 513/370 W
W×D×H 145×358×465 mm
Weight 16 kg