Heating circulators, HBC series
Supplier: IKA
4127000 4137000 4125000 4135000
IKAA4127000EA 13310 SGD
IKAA4127000 IKAA4137000 IKAA4125000 IKAA4135000
Heating circulators, HBC series
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
The HBC circulating baths have a high-quality stainless steel bath, an extended temperature range for tempering fluids in open and covered water baths up to 200/250 °C. The LED/ graphic TFT Display illustrates all relevant process parameters. The large surface of the radiator allows for an optimum heat exchange and a smooth warming of the medium. Units feature push and turn buttons for intuitive menu navigation. All models conform to the highest safety classification III (FL) for use with flammable liquids according to DIN 12876. HBC immersion circulators ensure short heating up times andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable