Magnetic hotplate stirrers, C-MAG HS digital series
Supplier: IKA
4240203 3487002 3487003 4240402 4240403 4240202
IKAA4240203EA 0 SGD
IKAA4240203 IKAA3487002 IKAA3487003 IKAA4240402 IKAA4240403 IKAA4240202
Magnetic hotplate stirrers, C-MAG HS digital series
Stirrers Magnetic Hotplate Stirrers
Compact digital magnetic stirrers with a chemically resistant ceramic top offer precise temperature control. The elevated control panel minimises contact with spills and enables easy reading and adjustment of the temperature. Units feature three operating modes: Standard, safe and adjustment protection. Stirrers have a digital error code display, a 'hot' top indicator to warn user that surface is hot and the strong magnetic adhesion prevents stirring bar decoupling.
  • Microprocessor control sends feedback to the motor and hotplate to maintain constant speed
More Product Information