Magnetic hotplate stirrers, RH basic and RH digital
Supplier: IKA
5019802 5019803 4678002 5019702 5019703 5029702 5029703
IKAA5019802EA 2290 SGD
IKAA5019802 IKAA5019803 IKAA4678002 IKAA5019702 IKAA5019703 IKAA5029702 IKAA5029703
Magnetic hotplate stirrers, RH basic and RH digital
Stirrers Magnetic Hotplate Stirrers
Magnetic hotplate stirrers with stainless steel heating plate, white units have an enamelled plate. All models are easy to operate and have adjustable safety temperature from 50 to 370 °C. High heat output of 600 watts and steel-copper compound hot plate ensures rapid heating.
  • RH basic includes M 10 thread for a support rod
  • Soft-start of the stirring motor for both models
  • Strong magnetic field and wide speed range for volumes up to 15 litres
  • RH digital has digital display of temperature and speed
  • Error code display and touch keypad (RH digital)