Magnetic hotplate stirrers, RET control-visc
Supplier: IKA
5020002
IKAA5020002EA 5240 SGD
IKAA5020002
Magnetic hotplate stirrers, RET control-visc
Stirrers Magnetic Hotplate Stirrers
RET control-visc safety magnetic stirrers with heating and integrated balance which are suitable for unsupervised operation. Clear, multilingual TFT display makes it easy to set all parameters and the sealed casing ensures a long service life. Integrated temperature control feature enables connection of a temperature probe, placed directly in the medium, to control medium temperature with a high degree of precision. The stainless steel composite hot plate (white units have a chemically resistant enamelled plate), reaches a temperature of 340 °C, enabling rapid heating. RS232 and USBMore Product Information