Decomposition system, AOD1
Supplier: IKA
8801300
IKAA8801300EA 0 SGD
IKAA8801300
Decomposition system, AOD1
Calorimeters
AOD 1 is used for oxidative decomposition of organic samples under pressure in a closed system.
  • Quantitative decomposition of all halogens, sulfur, as well as volatile metals e.g. As and Hg
  • Absorption of the combustion products in an aqueous medium
  • Catalytic support of the oxidation process with auto-regenerating catalytic surfaces
Pressure vessel of high-grade stainless steel.
Decomposition temperature up to 1200 °C
Maximum operating pressure during decomposition: 195 bar
Decomposition time <3 minutes
The decomposition vessel can be changedMore Product Information