Calorimeters, C 200
Supplier: IKA
8802500
IKAA8802500EA 0 SGD
IKAA8802500
Calorimeters, C 200
Calorimeters
Compact and space-saving semi automated calorimeter to determine calorific values of liquid and solid samples.
  • Manual oxygen filling and venting, manual water preparation
  • The C 5010 decomposition vessel can be equipped to use C 14 disposable crucible
  • Halogen resistant vessel for quantitative decomposition of halogens and sulphur
  • Includes external power pack, complies with all global voltages from 100 to 240 V AC, 50/60 Hz; calorimeter is powered with a low operating voltage of 24 V DC
  • Suitable for use with Calwin C 5040 calorimeter software: monitoring and
More Product Information