Single channel pipettes, mechanical, BagPipet®
Supplier: INTERSCIENCE
251091
INSC251091EA 903 SGD
INSC251091
Single channel pipettes, mechanical, BagPipet®
Pipettes Micropipettes
BagPipet® is an autoclavable pipette with three selectable volumes (0,1 ml / 0,9 ml / 1 ml). It is used with BagTips®, a single-use sterile and rigid straw.
  • Economical
  • No risk of cross-contamination
  • Light and ergonomic
  • Autoclavable and calibrated according to the GLP