Spiral plater/diluter, easySpiral Dilute®
Supplier: INTERSCIENCE
414000
INSC414000EA 0 SGD
INSC414000
Spiral plater/diluter, easySpiral Dilute®
Spiral Platers
easySpiral Dilute® is a 2-in-1 equipment: automatic diluter and plater. It allows to do 5 x serial 1/10 dilutions and automatic plating on 1 single Petri dish, with a countable range of 30 to 10¹² CFU/ml.Controlled by microprocessor it is possible to use Petri dishes with diameter: 55, 90 and 150 mm.With sample mixing before dilution/plating and monthly cleaning program. Successive plating capacity with the same sample: 20 Petri dishes (50 μl).
  • One-touch panel for routine serial dilutions and plating
  • Make as many dilutions as you want
  • Fast: disinfection, 5
More Product Information