Closures for test tubes and flasks
Supplier: JAECE INDUSTRIES
L800-D
JAECL800-DEA 127 SGD
JAECL800-D
Closures for test tubes and flasks
Closures Test/Sample Tube Closures
Identy-plug.
Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.
Cotton cheesecloth.
100% pure cotton, very absorbent and low linting; 28×24 thread count.
Bleached cotton balls.
Available large or medium.