Immersion thermostats, CORIO™ C, CD and CP
Supplier: JULABO GmbH
9011000 9012000
JULA9011000EA 2620 SGD
JULA9011000 JULA9012000
Immersion thermostats, CORIO™ C, CD and CP
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision, and a wide working temperature range. The CORIO™ C immersion thermostat is the basic model, it is ideal for standard internal applications, the CORIO™ CD/CP models are more advanced and can be equipped with an optional pump set for temperature control of external applications. The jet nozzle allows continuous adjustment of the pump stream in the bath. The bath attachment clamp enables either circulator to be mounted on virtually any bath (rectangular or round tanks).
Order Now
SPECIFICATIONS
Environmentally Preferable