Baths and circulators, stainless steel, CORIO™ C-B and CD-B series
Supplier: JULABO GmbH
9012419 9011405 9011413 9011417 9011419 9011427 9012405 9012413 9012417 9012427 9012433 9012439
JULA9012419EA 5740 SGD
JULA9012419 JULA9011405 JULA9011413 JULA9011417 JULA9011419 JULA9011427 JULA9012405 JULA9012413 JULA9012417 JULA9012427 JULA9012433 JULA9012439
Baths and circulators, stainless steel, CORIO™ C-B and CD-B series
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision and a wide working temperature range. The CORIO™ C immersion circulator is the basic model, it is ideal for standard internal applications, the CD model is more advanced and can used for temperature control of external applications if required. The jet nozzle allows continuous adjustment of the pump stream in the bath. The stainless steel baths are durable and have an integrated drain tap (except B5). These open heating bath circulators are ideal for temperature control of samples,More Product Information