Ultra-low temperature refrigerated circulators
Supplier: JULABO GmbH
9352755 9352752 9352756 9352793 9352791 9352790 9312681 9162681 9352755N150 9352752N150 9352795N 9352790N150
JULA9352755EA 92690 SGD
JULA9352755 JULA9352752 JULA9352756 JULA9352793 JULA9352791 JULA9352790 JULA9312681 JULA9162681 JULA9352755N150 JULA9352752N150 JULA9352795N JULA9352790N150
Ultra-low temperature refrigerated circulators
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Ultra-low temperature refrigerated circulators for heating and cooling. The units are suitable for external temperature applications with simultaneous operation in the bath tank. The units provide particularly high heating and cooling capacities for rapid heating and cooling times even for large volume, external systems. FP models offer proportional cooling control to ensure energy saving performance and to reduce heat generation to the environment. Models with the designation W are water cooled. All models have handles and castors which allow easy transportation and drain ports forMore Product Information