Cryo-Compact circulators, CF series
Supplier: JULABO GmbH
9400330 9400331 9400341 9400340
JULA9400330EA 17860 SGD
JULA9400330 JULA9400331 JULA9400341 JULA9400340
Cryo-Compact circulators, CF series
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Compact refrigerated/heating circulators which can be used on benchtop or in a fume hood, ideal for temperature control of distillation apparatus or mini plant installation. All four models feature pump connections for external temperature applications and a bath opening for temperature control of small objects directly in the circulator bath. The maximum ambient temperature of +40 °C and air cooled ventilation means the units can be positioned alongside other units or directly in a fume hood.
CF30/CF40 models are ideal for routine applications:
Working temperatures up to +150 °C withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Water Efficient