Temperature control systems, PRESTO®
Supplier: JULABO GmbH
9421912 9421923 9421913 9421922 9421912.TT 9421913.TT 9421922.TT 9421923.TT 9420452 9420452.T 9420852 9420852.T 9421852 9421852.T 9421502.T 9421502
JULA9421912EA 0 SGD
JULA9421912 JULA9421923 JULA9421913 JULA9421922 JULA9421912.TT JULA9421913.TT JULA9421922.TT JULA9421923.TT JULA9420452 JULA9420452.T JULA9420852 JULA9420852.T JULA9421852 JULA9421852.T JULA9421502.T JULA9421502
Temperature control systems, PRESTO®
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
PRESTO® systems cover a working temperature range of −93 to +250 °C with high cooling and heating capacity. These instruments can compensate for exothermic and endothermic reactions with extraordinary speed, they feature high flow rates, constant pressure and a controlled build-up of pump pressure. Changes in the temperature control of liquid viscosity are balanced dynamically. Permanent internal monitoring and self-lubricating pumps contribute to long service life. The integrated touch panel gives the user a clear view of important information. Fingertip control via the touch screen.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable