Water baths, PURA™
Supplier: JULABO GmbH
9550510 9550514
JULA9550510EA 3100 SGD
JULA9550510 JULA9550514
Water baths, PURA™
Baths Water Baths, Unstirred
PURA stainless steel water baths are functional and reliable for everyday operations due to their high material and component quality. The easy to use PURA water baths offer a number of functions and features that enable efficient use in the laboratory.
  • Intuitive, straightforward menu and bright, easy to read LED display
  • Splash-proof protected power switch
  • Integral drain screw (except PURA 4) and integrated timer
  • Built-in dry running protection
  • No interfering elements in the bath
  • Optical and acoustic alarm
  • Non slip rubber feet and ergonomic recessed grips
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable