Recirculating coolers, FC series
Supplier: JULABO GmbH
9600063 9600123 9600126 9600160 9600060 9600120 9600163 9600166 9601256
JULA9600063EA 29410 SGD
JULA9600063 JULA9600123 JULA9600126 JULA9600160 JULA9600060 JULA9600120 JULA9600163 JULA9600166 JULA9601256
Recirculating coolers, FC series
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Environmentally friendly recirculating coolers that save tap water, units are rapid and powerful, and offer short cool-down times and high efficiency. Due to the compact design, the coolers fit either on or under laboratory benchtops. FC units feature two separate multi-displays (LED) for actual and set point values.
  • Built-in heating element to allow cooling and heating applications with FC models
  • FC coolers have an adjustable ratio for inlet/outlet temperature
  • Connection for external PT100 sensor for external measurement and control (T-type models)
  • RS232
More Product Information