Recirculating coolers, F series
Supplier: JULABO GmbH
9620025
JULA9620025EA 8920 SGD
JULA9620025
Recirculating coolers, F series
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Environmentally friendly recirculating coolers that saves tap water, these units were designed for simple cooling tasks where reliable continuous operation is important. Due to their compact design, the coolers can be positioned either on or under laboratory bench tops. Units can be used with water, water/glycol or Julabo Thermal G. They are ideal for cooling rotary evaporators, Kjeldahl instruments, measurement cells, automated analysis systems, CCD cameras, polarimeters, refractometers, calorimeters and Soxhlet apparatus.
  • Environmentally friendly, units help to save costly tap water
More Product Information