Recirculating coolers, FL series
Supplier: JULABO GmbH
9661006 9661012 9663012 9660003 9661017 9663017 9663025 9663040 9666025 9666040 9666070 9666110 9671017 9673017 9673025 9673040 9676025 9676040 9676070 9676110
JULA9661006EA 18410 SGD
JULA9661006 JULA9661012 JULA9663012 JULA9660003 JULA9661017 JULA9663017 JULA9663025 JULA9663040 JULA9666025 JULA9666040 JULA9666070 JULA9666110 JULA9671017 JULA9673017 JULA9673025 JULA9673040 JULA9676025 JULA9676040 JULA9676070 JULA9676110
Recirculating coolers, FL series
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Compact, ergonomically designed chillers which have integrated stainless steel tanks with large volumes. Models can be either air or water cooled. Applications include use with rotary evaporators, autoclaves, reaction vessels and for routine laboratory cooling. All units have an LED temperature display for actual/set point integrated into a splash-proof keypad and recessed grips for easy relocation integrated in the housing. A hinged tray serves as a compartment for the operating manual and other documents. The removable venting grid allows for easy cleaning of the condenser. Venting slotsMore Product Information