Transparent bath tanks, polycarbonate, for CORIO C / CD
Supplier: JULABO GmbH
9901305 9901309 9901319 9901327
JULA9901305EA 694 SGD
JULA9901305 JULA9901309 JULA9901319 JULA9901327
Transparent bath tanks, polycarbonate, for CORIO C / CD
Baths Water Baths, Unstirred
Transparent PC bath tanks in different sizes. For use with all CORIO C and CD circulators.
Temperature range: +20...+100 °C
Order Now
SPECIFICATIONS
Environmentally Preferable