Bath tanks, stainless steel, for CORIO C / CD
Supplier: JULABO GmbH
9903405 9903413 9903417 9903419 9903427 9903433 9903439
JULA9903405EA 1000 SGD
JULA9903405 JULA9903413 JULA9903417 JULA9903419 JULA9903427 JULA9903433 JULA9903439
Bath tanks, stainless steel, for CORIO C / CD
Baths Water Baths, Unstirred
Stainless steel bath tanks in different sizes. For use with all CORIO C and CD circulators.
Temperature range: +20...+150 °C
Order Now
SPECIFICATIONS
Environmentally Preferable