Safety Funnel, for drums
Supplier: JUSTRITE MANUFACTURING
08201 08203 08207 08205 08208
JUSU08201EA 300 SGD
JUSU08201 JUSU08203 JUSU08207 JUSU08205 JUSU08208
Safety Funnel, for drums
Funnels Safety Funnels
Galvanised steel, 18-gauge, with electrostatically applied epoxy powder coat paint.
  • Wide mouth opening, stay-opened lid
  • Self-closing lin incase of fire
  • Built-in flame arrester on most models to prevent fire intrusion
  • Funnels for use with viscous liquids are fitted with brass tube
These safety drum funnels are mainly used to prevent spills while transferring flammable solvents, thinners, oils or paint wastes.
Approved by FM and compliance with EPA and OSHA regulations.
Order Now
SPECIFICATIONS
Environmentally Preferable