Bench cans for solvents
Supplier: JUSTRITE MANUFACTURING
10175 10295 10370 10375 10575 10775
JUSU10175EA 164 SGD
JUSU10175 JUSU10295 JUSU10370 JUSU10375 JUSU10575 JUSU10775
Bench cans for solvents
Safety Cans
These bench cans for solvent safety have plated steel dasher which work as an effective flame arrester to reduce the risk of fire flashback. Hinged cover minimises evaporation losses and protected rim reduces the possibility of hand injury.
  • Sturdy 24-gauge steel body construction with tough powder-coat finish
  • Compliance with FM and TUV approved
Order Now
SPECIFICATIONS
Environmentally Preferable