Safety drip cans
Supplier: JUSTRITE MANUFACTURING
10376
JUSU10376EA 187 SGD
JUSU10376
Safety drip cans
Safety Cans
These steel drip cans are used ideally with flammable liquids. Constructed of 24-gauge coated steel with a durable powder paint finish and perforated metal fire baffle, which acts as a flame arrester to protect contents from fire exposure. They have side handles for easy portability.