Safety Bottle Jacket
Supplier: JUSTRITE MANUFACTURING
12009
JUSU12009EA 58.8 SGD
JUSU12009
Safety Bottle Jacket
Carriers Bottle Carriers/Containers
PE, translucent
  • Translucent colour aids in detecting liquid level
  • Jacket acts as a shock absorber, minimising breakage and spills due to bumps or falls
Bottles fit into the bottom half of the jacket and lid twists on securely for safe pouring.