Safety dispensing cans, HDPE
Supplier: JUSTRITE MANUFACTURING
14002 14004 14010
JUSU14002EA 144 SGD
JUSU14002 JUSU14004 JUSU14010
Safety dispensing cans, HDPE
Safety Cans
These FM approved brass valves are ideal for use with flammables.
HDPE, textured.
  • Permit easy, one-hand dispensing of liquids directly onto work surface
  • Leakproof, self-closing, brass dispenser valve minimises spills and controls excess