Lockers for LPG and compressed gas cylinders
Supplier: JUSTRITE MANUFACTURING
23001 23002 23003 23004 23005 23006 23007 23008 23009 23010 23011
JUSU23001EA 3120 SGD
JUSU23001 JUSU23002 JUSU23003 JUSU23004 JUSU23005 JUSU23006 JUSU23007 JUSU23008 JUSU23009 JUSU23010 JUSU23011
Lockers for LPG and compressed gas cylinders
Furniture Lockers
These durable cylinder lockers made of welded aluminum, 1,65 mm thick metal panels, are ideal for storing LPG and compressed gas cylinders either horizontally or vertically.
  • Easy to use
  • Rust, corrosion and tamper resistant
  • For 9 or 15 kg cylinders
Welded security lug on front panel door accepts a padlock for extra protection, also the heavier gauge aluminum mesh and extruded frame add rigidity to the locker. An exclusive solid aluminium roof panel that slants toward the back help to shed water and ice.
Certifications: Compliance with NFPA and OSHA approved
Order Now
SPECIFICATIONS
Environmentally Preferable