Waste receptacle, Heavy-Duty, Cease-Fire®
Supplier: JUSTRITE MANUFACTURING
26430
JUSU26430EA 636 SGD
JUSU26430
Waste receptacle, Heavy-Duty, Cease-Fire®
Bins Waste Bins
These waste receptacles with steel heads are specially designed to direct smoke and gas from any fire inside the container, back into the combustion area, cutting off the air supply and extinguishing flames in seconds. UL listed and FM approved.
  • Provide around-the-clock protection against trash fires
  • Drums should be filled 75% full only