Smoking receptacle, Cease Fire®
Supplier: JUSTRITE MANUFACTURING
26800
JUSU26800EA 187 SGD
JUSU26800
Smoking receptacle, Cease Fire®
Bins Waste Bins
The original smoker’s Cease Fire® cigarette butt receptacle’s self-extinguishing design safely collects unsightly cigarette litter and reduces the risk of fire. Unique rain bonnet opening keeps rain out and discourages other trash. Has a big four gallon galvanised steel pail inside. 'Drip-lip' internal construction prevents tar condensation from escaping, keeping exterior clean from dark, messy drip marks.
  • Made of flame-retardant, sturdy polyethylene
  • Tie-down notches offer security and greater stability under severe weather conditions
  • Fire tested and
More Product Information