Diverters for ceiling leaks
Supplier: JUSTRITE MANUFACTURING
28300 28302 28308 28310 28314
JUSU28300EA 206 SGD
JUSU28300 JUSU28302 JUSU28308 JUSU28310 JUSU28314
Diverters for ceiling leaks
Spill Control Materials
These ceiling leak diverters redirect roof or pipe leaks to prevent from water damages. The durable, puncture resistant and watertight PVC-coated yellow fabric is designed to channel overhead pipe and ceiling leaks to containers or floor drains.
  • Available with high strength magnets for steel rafter attachment, or grommets for bungee or zip tie attachment
  • Constructed with cinched corners for quick and easy installation
  • Simply connect to a standard garden hose to drain water safely away
Flow rate: 12,5 L/min
Order Now
SPECIFICATIONS
Environmentally Preferable