Spill containment pop-up pools
Supplier: JUSTRITE MANUFACTURING
28319 28321 28323 28325
JUSU28319EA 314 SGD
JUSU28319 JUSU28321 JUSU28323 JUSU28325
Spill containment pop-up pools
Spill Control Materials
The space-saving and lightweight pools are designed to collect spills from punctured saddle tanks, or leaking drums, hydraulic lines, pipes, and machinery. The durable PVC coated vinyl fabric is 20% heavier than other models, providing increased abrasion resistance and reliable seam adhesions.
  • Folds tightly and stows easily
  • Yellow colour provides visibility and easy identification
  • Embedded foam ring top raises containment wall as fluid level increases
Certifications: Helps to comply with EPA for containment and spill prevention (SPCC).